missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Hepatitis A HAV P2C-P3B Protein

A cDNA sequence encoding the HAV P2C-P3B was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12109-1mg

 View more versions of this product

Product Code. 15949398

  • £1320.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Hepatitis A HAV P2C-P3B Protein
Research Use Only
Recombinant Protein
E. coli
10mM Tris-HCl, pH 9.6, 0.1% SDS and 50% glycerol.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Hepatitis A
GST
HAV P2C-P3B protein is >90% pure as determined by 10% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers