missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis A HAV P2C-P3B Protein
A cDNA sequence encoding the HAV P2C-P3B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12109-100ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis A HAV P2C-P3B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
10mM Tris-HCl, pH 9.6, 0.1% SDS and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis A | |
GST | |
HAV P2C-P3B protein is >90% pure as determined by 10% PAGE (coomassie staining). |