missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant H. pylori DNA protection during starvation Protein

Product Code. 16077610
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16077610

Brand: enQuireBio™ QP7186ec50ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Helicobacter pylori (strain ATCC 700392 / 26695) (Campylobacter pylori) DNA protection during starvation protein, was expressed in the hosts and tags indicated. option, abo

Specifications

Accession Number NP_207041.1
Gene ID (Entrez) 898765
Name DNA protection during starvation Protein
Quantity 50 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol dps
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity H. pylori
Species E. coli
Protein Tag His-SUMO
Sequence MKTFEILKHL QADAIVLFMK VHNFHWNVKG TDFFNVHKAT EEIYEEFADM FDDLAERIVQ LGHHPLVTLS EAIKLTRVKE ETKTSFHSKD IFKEILEDYK YLEKEFKELS NTAEKEGDKV TVTYADDQLA KLQKSIWMLQ AHLA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.