missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Guinea Pig PSAP / Prosaposin Protein
A DNA sequence encoding the Cavia porcellus (Guinea pig) PSAP / Prosaposin, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9351-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
PSAP / Prosaposin Protein | |
Research Use Only | |
PSAP | |
Recombinant Protein | |
Yeast | |
ESVTCKACEY VVKKVMELID NNRTEEKIIH ALDSVCALLP ESVSEVCQEV VDTYGDSIVA LLLQEMSPEL VCSELGLCMS G | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Guinea Pig | |
His | |
Tris-based buffer, 50% glycerol |