missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Guinea Pig CD46 Protein

A DNA sequence encoding the Cavia porcellus (Guinea pig) CD46, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5789-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15927769

  • £715.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001166399.2
CD46 Protein
Research Use Only
Cd46
Recombinant Protein
E. coli
CVLPPPFEAM EPINPKPYYE IGEKVEYRCK KGYLRQPFYL MVATCEKNHS WVPITDDGCI KKQCTYLNPP PKGRVEYING TRTWGDIVHF SCVEGFYVSG IAALSCELRG DNVDWNGRVP TCEKVLCSPP PKIQNGKYTF SDVQVFEYFE AVTYSCDAVQ GPDKLSLVGN EVLYCAGHQK WSSAAPECKV VKCPLPVVKN GKQISGLGQT FFYQATVTFQ CLPGFYFNGS STVVCGSDNT WKPSIPECLK GPKPTHPTKP PVYNYPGYPN PREGIFDQEL N
Greater than 90% as determined by SDS-PAGE.
100135498
100 μg
Not determined.
Extracellular Domain
Guinea Pig
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers