missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant GFP/SNAP25B/VAMP-2 Protein
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant GFP/SNAP25B/VAMP-2 Protein is derived from E. coli. The Recombinant GFP/SNAP25B/VAMP-2 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
| Formulation | Supplied as a 0.2μm filtered solution in Tris, NaCl, DTT and Tween™. |
| Molecular Weight (g/mol) | 52 kDa |
| Name | GFP/SNAP25B/VAMP-2 |
| Quantity | 50μg |
| Source | E. coli-derived GFP/SNAP25B/VAMP-2 protein, (N-terminus) MVKSADI 10-His tag Linker 1 GFP (Ser2-Lys238) Accession &Num; ACS44347 Linker 2 Human SNAP25B (Asn93-Gly206) Accession &Num; P60880 Human VAMP-2 (Ser2-Lys94) Accession &Num; P63027 Cys (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | >85%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction