missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Feline IL-12/IL-23 p40 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£353.12
Specifications
Gene ID (Entrez) | 493741 |
---|---|
Molecular Weight (g/mol) | 36 kDa |
Name | IL-12/IL-23 p40 |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived feline IL-12/IL-23 p40 protein Ile23-Ser329 (Glu167Gly),with a C-terminal 10-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18187220
|
R&D Systems™
2117-FL-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£353.12
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18138320
|
R&D Systems™
2117-FL-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£353.12
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Feline IL-12/IL-23 p40 Protein is derived from NS0. The Recombinant Feline IL-12/IL-23 p40 Protein has been validated for the following applications: Binding Activity.Specifications
493741 | |
IL-12/IL-23 p40 | |
Mouse myeloma cell line,NS0-derived feline IL-12/IL-23 p40 protein Ile23-Ser329 (Glu167Gly),with a C-terminal 10-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
36 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |