missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Equine IL-10 Protein
Description
The Recombinant Equine IL-10 Protein is derived from E. coli. The Recombinant Equine IL-10 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Molecular Weight (g/mol) | 19 kDa |
| Name | IL-10 |
| Quantity | 10 μg |
| Source | E. coli-derived equine IL-10 protein Ser19-Asn178 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
| Structural Form | Noncovalently-linked homodimer |
| Gene Alias | CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10MGC126451, interleukin 10, interleukin-10, TGIF |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?