missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Equine IFN-gamma Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£341.24
Specifications
Molecular Weight (g/mol) | 16.7 kDa |
---|---|
Name | IFN-gamma |
Quantity | 25μg |
Source | E. coli-derived equine IFN-gamma protein Ala25-Gln166 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18198240
|
R&D Systems™
1586-HG-025 |
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80 with BSA as a carrier protein. |
£341.24
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18107140
|
R&D Systems™
1586-HG-025/CF |
Lyophilized from a 0.2μm filtered solution in Sodium Succinate, Mannitol and Tween™ 80. |
£341.24
25µg |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Equine IFN-gamma Protein is derived from E. coli. The Recombinant Equine IFN-gamma Protein has been validated for the following applications: Bioactivity.Specifications
16.7 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
IFN-gamma | |
E. coli-derived equine IFN-gamma protein Ala25-Gln166 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |