missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Epstein-Barr Latent membrane protein 1
A DNA sequence encoding the Epstein-Barr virus (strain Raji) (HHV-4) (Human herpesvirus 4) Latent membrane protein 1, was expressed in the hosts and tags indicated.
£321.05 - £2888.42
Specifications
Name | Latent membrane protein 1 |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | LMP1 |
Protein Length | Cytoplasmic Domain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16087090
|
enQuireBio™
QP7024-YE-100UG |
100 μg |
£800.00
100µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16027100
|
enQuireBio™
QP7024-YE-500UG |
500 μg |
£1930.00
500µg |
Estimated Shipment: 27-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16097090
|
enQuireBio™
QP7024-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16017100
|
enQuireBio™
QP7024-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16037100
|
enQuireBio™
QP7024-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16007100
|
enQuireBio™
QP7024-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Latent membrane protein 1 | |
Not determined. | |
Cytoplasmic Domain | |
Epstein-Barr | |
His | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
LMP1 | |
Recombinant Protein | |
Yeast | |
YYHGQRHSDE HHHDDSLPHP QQATDDSSNQ SDSNSNEGRH LLLVSGAGDG PPLCSQNLGA PGGGPNNGPQ DPDNTDDNGP QDPDNTDDNG PHDPLPQDPD NTDDNGPQDP DNTDDNGPHD PLPHNPSDSA GNDGGPPQLT EEVENKGGDQ GPPLMTDGGG GHSHDSGHDG IDPHLPTLLL GTSGSGGDDD DPHGPVQLSY YD | |
Greater than 90% as determined by SDS-PAGE. |