missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Epstein-Barr Envelope glycoprotein GP350 Protein

Product Code. 16018380
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16018380

Brand: enQuireBio™ QP7351ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Epstein-Barr virus (strain B95-8) (HHV-4) (Human herpesvirus 4) Envelope glycoprotein GP350, was expressed in the hosts and tags indicated.

Specifications

Accession Number YP_401667.1
Gene ID (Entrez) 3783713
Name Envelope glycoprotein GP350 Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol BLLF1
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Epstein-Barr
Species E. coli
Protein Tag His-SUMO
Sequence MEAALLVCQY TIQSLIHLTG EDPGFFNVEI PEFPFYPTCN VCTADVNVTI NFDVGGKKHQ LDLDFGQLTP HTKAVYQPRG AFGGSENATN LFLLELLGAG ELALTMRSKK LPINVTTGEE QQVSLESVDV YFQDVFGTMW CHHAEMQNPV YLIPETVPYI KWDNCNSTNI TAVVRAQGLD VTLPLSLPTS AQDSNFSVKT EMLGNEIDIE CIMEDGEISQ VLPGDNKFNI TCSGYESHVP SGGILTSTSP VATPIPGTGY AYSLRLTPRP VSRFLGNNSI LYVFYSGNGP KASGGDYCIQ SNIVFSDEIP ASQDMPTNTT DITYVGDNAT YSVPMVTSED ANSPNVTVTA FWAWPNNTET DFKCKWTLTS GTPSGCENIS GAFASNRTFD ITVSGLGTAP KTLIITRTAT NATTTTHKVI FSKAPESTTT SPTLNTTGFA DPNTTTGLPS STHVPTNLTA PASTGPTVST ADVTSPTPAG TTSGASPVTP SPS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 80% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.