missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant eGFP Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
Specifications
Specifications
| Accession Number | P42212 |
| Concentration | LYOPH |
| For Use With (Application) | Western Blot, PAGE |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Molecular Weight (g/mol) | 26.9 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/ug of eGFP as determined by LAL method. |
| Gene Alias | Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction