missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant eGFP Protein

Product Code. 18420902 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
Unit Size:
10µg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420902 10 μg 10µg
18444362 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18420902 Supplier Novus Biologicals™ Supplier No. NBP23492310ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42212
Concentration LYOPH
For Use With (Application) Western Blot, PAGE
Formulation Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4.
Molecular Weight (g/mol) 26.9 kDa
Quantity 10 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of eGFP as determined by LAL method.
Gene Alias Enhanced Green Fluorescent Protein, GFP, Green Fluorescent Protein
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Inhibitor Properties Use in WB reported in scientific literature (PMID:10.1007/s00299-021-02700-w)
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein eGFP
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.