missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant EBOV GP (Mucin Domain Deleted) Protein, CF
Description
Measured by its binding ability in a functional ELISA. When Recombinant Viral EBOV GP (mucin domain deleted, Catalog # 10585-EB) is immobilized at 2 μg/mL (100 μL/well), the concentration of Recombinant Human CLEC10A/CD301 (Catalog # 4888-CL ) binds with an ED50 of 1-10 ng/mL.
Specifications
Specifications
| Accession Number | NP_0066246.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 3160774 |
| Molecular Weight (g/mol) | M.W. (Observed): 63-75 kDa, under non-reducing conditions, M.W. (Theoretical): 51 kDa |
| Quantity | 100 μg |
| Source | Human embryonic kidney cell, HEK293-derived viral EBOV GP protein Ebola Virus GP1 (Ile33-Val311) (Thr42Val, Thr230Val) (N-treminus) Accession # NP_0066246.1 Ebola Virus GP2 (Thr464-Asp632) Accession # NP_066246.1 HHHHHH (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | bovGP, second secreted glycoprotein, small secreted glycoprotein, Spike glycoprotein |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?