missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli TPX Protein
A cDNA sequence encoding the TPX was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13801-20ug
Additional Details : Weight : 0.01000kg
Specifications
TPX Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MSQTVHFQGN PVTVANSIPQ AGSKAQTFTL VAKDLSDVTL GQFAGKRKVL NIFPSIDTGV CAASVRKFNQ LATEIDNTVV LCISADLPFA QSRFCGAEGL NNVITLSTFR NAEFLQAYGV AIADGPLKGL AARAVVVIDE NDNVIFSQLV DEITTEPDYE AALAVLKA | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
E. coli | |
His | |
The Recombinant TPX solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) 10% glycerol, 2mM DTT and 0.1M NaCl. |