missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli tetR Protein

Product Code. 16008770
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16008770

Brand: enQuireBio™ QP7416ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli tetR, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_000088605.1
Gene ID (Entrez) 31515962
Name tetR Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol tetR
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence MSRLDKSKVI NSALELLNEV GIEGLTTRKL AQKLGVEQPT LYWHVKNKRA LLDALAIEML DRHHTHFCPL EGESWQDFLR NNAKSFRCAL LSHRDGAKVH LGTRPTEKQY ETLENQLAFL CQQGFSLENA LYALSAVGHF TLGCVLEDQE HQVAKEERET PTTDSMPPLL RQAIELFDHQ GAEPAFLFGL ELIICGLEKQ LKCESGS
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado