missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Signal peptidase I Protein
A DNA sequence encoding the Escherichia coli (strain K12) Signal peptidase I, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7340-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417063.1 | |
Signal peptidase I Protein | |
Research Use Only | |
lepB | |
Recombinant Protein | |
E. coli | |
RSFIYEPFQI PSGSMMPTLL IGDFILVEKF AYGIKDPIYQ KTLIETGHPK RGDIVVFKYP EDPKLDYIKR AVGLPGDKVT YDPVSKELTI QPGCSSGQAC ENALPVTYSN VEPSDFVQTF SRRNGGEATS GFFEVPKNET KENGIRLSER KETLGDVTHR ILTVPIAQDQ VGMYYQQPGQ QLATWIVPPG QYFMMGDNRD NSADSRYWGF VPEANLVGRA TAIWMSFDKQ EGEWPTGLRL SRIGGIH | |
Greater than 90% as determined by SDS-PAGE. |
947040 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |