missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant E. coli RNase P protein component Protein
Shop All Bio Techne ProductsDescription
Amino Acid Sequence: MVKLAFPREL RLLTPSQFTF VFQQPQRAGT PQITILGRLN SLGHPRIGLT VAKKNVRRAH ERNRIKRLTR ESFRLRQHEL PAMDFVVVAK KGVADLDNRA LSEALEKLWR RHCRLARGS
Specifications
Specifications
| Accession Number | NP_418159.1 |
| Concentration | 0.25 mg/ml |
| For Use With (Application) | SDS-PAGE |
| Formulation | PBS (pH 7.4), 10% glycerol |
| Gene ID (Entrez) | 948215 |
| Molecular Weight (g/mol) | M.W. theroretical: 13.7 kDa |
| Quantity | 250 μg |
| Source | E. Coli |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <:1 EU / 1 ug of protein (determined by LAL method) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?