missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Penicillin-binding protein 1A Protein
A DNA sequence encoding the Escherichia coli (strain K12) Penicillin-binding protein 1A, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_417855.4 |
---|---|
Gene ID (Entrez) | 947907 |
Name | Penicillin-binding protein 1A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16058700
|
enQuireBio™
QP7405-ec-10ug |
10 μg | |||||||
16098700
|
enQuireBio™
QP7405-ec-50ug |
50 μg | |||||||
16048700
|
enQuireBio™
QP7405-ec-100ug |
100 μg |
£715.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16078700
|
enQuireBio™
QP7405-ec-200ug |
200 μg | |||||||
16088700
|
enQuireBio™
QP7405-ec-500ug |
500 μg | |||||||
16068700
|
enQuireBio™
QP7405-ec-1mg |
1 mg |
£2480.00
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_417855.4 | |
Penicillin-binding protein 1A Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
947907 | |
Research Use Only | |
mrcA | |
Recombinant Protein | |
E. coli | |
MLDEGYITQQ QFDQTRTEAI NANYHAPEIA FSAPYLSEMV RQEMYNRYGE SAYEDGYRIY TTITRKVQQA AQQAVRNNVL DYDMRHGYRG PANVLWKVGE SAWDNNKITD TLKALPTYGP LLPAAVTSAN PQQATAMLAD GSTVALSMEG VRWARPYRSD TQQGPTPRKV TDVLQTGQQI WVRQVGDAWW LAQVPEVNSA LVSINPQNGA VMALVGGFDF NQSKFNRATQ ALRQVGSNIK PFLYTAAMDK GLTLASMLND VPISRWDASA GSDWQPKNSP PQYAGPIRLR QGLGQSKNVV M | |
Greater than 80% as determined by SDS-PAGE. |