missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Outer membrane protein 1
A DNA sequence encoding the Escherichia coli (strain K12) Outer membrane protein C, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7295-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_416719.1 | |
Outer membrane protein 1 | |
Research Use Only | |
ompC | |
Recombinant Protein | |
E. coli | |
AEVYNKDGNK LDLYGKVDGL HYFSDNKDVD GDQTYMRLGF KGETQVTDQL TGYGQWEYQI QGNSAENENN SWTRVAFAGL KFQDVGSFDY GRNYGVVYDV TSWTDVLPEF GGDTYGSDNF MQQRGNGFAT YRNTDFFGLV DGLNFAVQYQ GKNGNPSGEG FTSGVTNNGR DALRQNGDGV GGSITYDYEG FGIGGAISSS KRTDAQNTAA YIGNGDRAET YTGGLKYDAN NIYLAAQYTQ TYNATRVGSL GWANKAQNFE AVAQYQFDFG LRPSLAYLQS KGKNLGRGYD DEDILKYVDV GATYYFNKNM STYVDYKINL LDDNQFTRDA GINTDNIVAL GLVYQF | |
Greater than 80% as determined by SDS-PAGE. |
946716 | |
50 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |