missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Outer membrane protein 1

A DNA sequence encoding the Escherichia coli (strain K12) Outer membrane protein C, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7295-ec-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16048050

  • £292.00 / 10µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_416719.1
Outer membrane protein 1
Research Use Only
ompC
Recombinant Protein
E. coli
AEVYNKDGNK LDLYGKVDGL HYFSDNKDVD GDQTYMRLGF KGETQVTDQL TGYGQWEYQI QGNSAENENN SWTRVAFAGL KFQDVGSFDY GRNYGVVYDV TSWTDVLPEF GGDTYGSDNF MQQRGNGFAT YRNTDFFGLV DGLNFAVQYQ GKNGNPSGEG FTSGVTNNGR DALRQNGDGV GGSITYDYEG FGIGGAISSS KRTDAQNTAA YIGNGDRAET YTGGLKYDAN NIYLAAQYTQ TYNATRVGSL GWANKAQNFE AVAQYQFDFG LRPSLAYLQS KGKNLGRGYD DEDILKYVDV GATYYFNKNM STYVDYKINL LDDNQFTRDA GINTDNIVAL GLVYQF
Greater than 80% as determined by SDS-PAGE.
946716
10 μg
Not determined.
Full Length
E. coli
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers