missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli NTH Protein
A cDNA sequence encoding the NTH was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12892-1mg
Additional Details : Weight : 0.01000kg
Specifications
947122 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MNKAKRLEIL TRLRENNPHP TTELNFSSPF ELLIAVLLSA QATDVSVNKA TAKLYPVANT PAAMLELGVE GVKTYIKTIG LYNSKAENII KTCRILLEQH NGEVPEDRAA LEALPGVGRK TANVVLNTAF GWPTIAVDTH IFRVCNRTQF APGKNVEQVE EKLLKVVPAE FKVDCHHWLI LHGRYTCIAR KPRCGSCIIE DLCEYKEKVD I | |
Greater than 85.0% as determined by SDS-PAGE. |
NTH Protein | |
Research Use Only | |
nth | |
E. coli | |
His | |
The NTH solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 1mM DTT, 0.1mM PMSF and 40% glycerol. |