missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli nrdA Protein

Product Code. 16037930
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16037930

Brand: enQuireBio™ QP7262ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) nrdA, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_416737.1
Gene ID (Entrez) 946612
Name nrdA Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol nrdA
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence MNQNLLVTKR DGSTERINLD KIHRVLDWAA EGLHNVSISQ VELRSHIQFY DGIKTSDIHE TIIKAAADLI SRDAPDYQYL AARLAIFHLR KKAYGQFEPP ALYDHVVKMV EMGKYDNHLL EDYTEEEFKQ MDTFIDHDRD MTFSYAAVKQ LEGKYLVQNR VTGEIYESAQ FLYILVAACL FSNYPRETRL QYVKRFYDAV STFKISLPTP IMSGVRTPTR QFSSCVLIEC GDSLDSINAT SSAIVKYVSQ RAGIGINAGR IRALGSPIRG GEAFHTGCIP FYKHFQTAVK SCSQGGVRGG AATLFYPMWH LEVESLLVLK NNRGVEGNRV RHMDYGVQIN KLMYTRLLKG EDITLFSPSD VPGLYDAFFA DQEEFERLYT KYEKDDSIRK QRVKAVELFS LMMQERASTG RIYIQNVDHC NTHSPFDPAI APVRQSNLCL EIALPTKPLN DVNDENGEIA LCTLSAFNLG AINNLDELEE LAILAVRALD ALLDYQDYPI PAAKRGAMGR RTLGIGVINF AYYLAKHGKR YSDGSANNLT HKTFEAIQYY LLKASNELAK EQGACPWFNE TTYAKGILPI DTYKKDLDTI ANEPLHYDWE ALRESIKTHG LRNSTLSALM PSETSSQISN ATNGIEPPRG YVSIKASKDG ILRQVVPDYE HLHDAYELLW EMPGNDGYLQ LVGIMQKFID QSISANTNYD PSRFPSGKVP MQQLLKDLLT AYKFGVKTLY YQNTRDGAED AQDDLVPSIQ DDGCESGACK I
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.