missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli NAD(P) transhydrogenase subunit alpha Protein
A DNA sequence encoding the Escherichia coli (strain K12) NAD(P) transhydrogenase subunit alpha, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9674-ye-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_416120.1 | |
NAD(P) transhydrogenase subunit alpha Protein | |
Research Use Only | |
pntA | |
Recombinant Protein | |
Yeast | |
MRIGIPRERL TNETRVAATP KTVEQLLKLG FTVAVESGAG QLASFDDKAF VQAGAEIVEG NSVWQSEIIL KVNAPLDDEI ALLNPGTTLV SFIWPAQNPE LMQKLAERNV TVMAMDSVPR ISRAQSLDAL SSMANIAGYR AIVEAAHEFG RFFTGQITAA GKVPPAKVMV IGAGVAGLAA IGAANSLGAI VRAFDTRPEV KEQVQSMGAE FLELDFKEEA GSGDGYAKVM SDAFIKAEME LFAAQAKEVD IIVTTALIPG KPAPKLITRE MVDSMKAGSV IVDLAAQNGG NCEYTVPGEI FTTENGVKVI GYTDLPGRLP TQSSQLYGTN LVNLLKLLCK EKDGNITVDF DDVVIRGVTV IRAGEITWPA PPIQVSAQPQ AAQKAAPEVK TEEKCTCSPW R | |
Greater than 90% as determined by SDS-PAGE. |
946628 | |
1 mg | |
Not determined. | |
Cytoplasmic Domain | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |