missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant E. coli Methionine Aminopeptidase/METAP
Description
Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC (Catalog # ES017 ). The resulting GP-AMC is cleaved by Recombinant Human DPPIV/CD26 (Catalog # 9168-SE ). The specific activity is >0.5 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on www.RnDSystems.com .
Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC (Catalog # ES017 ). The resulting GP-AMC is cleaved by Recombinant Human DPPIV/CD26 (Catalog # 9168-SE ). The specific activity is >0.5 pmol/min/μg, as measured under the described conditions. See Activity Assay Protocol on www.RnDSystems.com .
Specifications
Specifications
| Accession Number | NP_414710 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. |
| Gene ID (Entrez) | 947882 |
| Molecular Weight (g/mol) | M.W. (Observed): 36 kDa, reducing conditions; M.W. (theoretical): 31 kDa |
| Name | Methionine Aminopeptidase |
| Quantity | 20 μg |
| Source | E. coli-derived e. coli Methionine Aminopeptidase protein Ala2-Glu264, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?