missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli GRXB Protein
A cDNA sequence encoding the GRXB was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12054-1mg
Additional Details : Weight : 0.01000kg
Specifications
946926 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MKLYIYDHCP YCLKARMIFG LKNIPVELHV LLNDDAETPT RMVGQKQVPI LQKDDSRYMP ESMDIVHYVD KLDGKPLLTG KRSPAIEEWL RKVNGYANKL LLPRFAKSAF DEFSTPAARK YFVDKKEASA GNFADLLAHS DGLIKNISDD LRALDKLIVK PNAVNGELSE DDIQLFPLLR NLTLVAGINW PSRVADYRDN MAKQTQINLL SSMAI | |
Greater than 95% as determined by SDS-PAGE. |
GRXB Protein | |
Research Use Only | |
grxB | |
E. coli | |
His | |
GRXB protein solution (1 mg/ml) 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 50mM NaCl. |