missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant E. coli Glycerol Kinase Protein
Click to view available options
:
20ug; Carrier Free
Description
Measured by its ability to transfer phosphate from ATP to glycerol. The specific activity is >1,500 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | AAC76908 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, NaCl, Glycerol, Brij-35 and DTT. |
| Gene ID (Entrez) | 2710 |
| Molecular Weight (g/mol) | M.W. (Observed): 52-55 kDa, reducing conditions; M.W. (theoretical): 57 kDa |
| Name | Glycerol Kinase |
| Quantity | 20 μg |
| Source | E. coli-derived e. coli Glycerol Kinase protein Met1-Glu502, with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction