missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Ferric uptake regulation Protein
A DNA sequence encoding the Escherichia coli (strain K12) Ferric uptake regulation protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7394-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_415209.1 | |
Ferric uptake regulation Protein | |
Research Use Only | |
fur | |
Recombinant Protein | |
Yeast | |
TDNNTALKKA GLKVTLPRLK ILEVLQEPDN HHVSAEDLYK RLIDMGEEIG LATVYRVLNQ FDDAGIVTRH NFEGGKSVFE LTQQHHHDHL ICLDCGKVIE FSDDSIEARQ REIAAKHGIR LTNHSLYLYG HCAEGDCRED EHAHEGK | |
Greater than 90% as determined by SDS-PAGE. |
945295 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |