missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant E. coli DsbG Protein
Shop All Bio Techne Products
Click to view available options
:
0.1mg; Unlabeled
Description
An un-tagged recombinant protein corresponding to the amino acids 18-248 of E.coli DsbG The Recombinant E. coli DsbG Protein is derived from E. coli. The Recombinant E. coli DsbG Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_415137 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0), 2mM EDTA, 10% glycerol |
| Gene ID (Entrez) | 945224 |
| Molecular Weight (g/mol) | 25.8kDa |
| Name | DsbG Protein |
| Purification Method | Affinity purified |
| Immunogen | MEELPAPVKA IEKQGITIIK TFDAPGGMKG YLGKYQDMGV TIYLTPDGKH AISGYMYNEK GENLSNTLIE KEIYAPAGRE MWQRMEQSHW LLDGKKDAPV IVYVFADPFC PYCKQFWQQA RPWVDSGKVQ LRTLLVGVIK PESPATAAAI LASKDPAKTW QQYEASGGKL KLNVPANVST EQMKVLSDNE KLMDDLGANV TPAIYYMSKE NTLQQAVGLP DQKTLNIIMG NK |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Purity or Quality Grade | >95%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction