missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant E. coli Double-Stranded Uracil-DNA Glycosylase His Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
- A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-168 of E.coli Double-Stranded Uracil-DNA Glycosylase
- G/U mismatch-specific DNA glycosylase, xanthine DNA glycosylase, also known as mug, belongs to the TDG/mug DNA glycosylase family.
- It has been proposed that the Mug protein excises 3, N4-ethenocytosine and removes the uracil base from mismatches in the order of U:G>U:A, although the biological role remains unclear.
- The enzyme Uracil-N-Glycosylase removes uracil from the DNA leaving an AP site.
- It is capable of hydrolyzing the carbon-nitrogen bond between the sugar-phosphate backbone of the DNA and the mispaired base.
- The complementary strand guanine functions in substrate recognition.
- Recombinant E.coli mug protein, fused to His-tag at N-terminus, was expressed in E.coli and purified by using conventional chromatography techniques.
- This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Specifications
Specifications
| Concentration | 0.5 mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20 mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol |
| Molecular Weight (g/mol) | 21.1 kDa |
| Quantity | 50 μg |
| Source | E. coli |
| Storage Requirements | Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | double-strand uracil-DNA glycosylase, Double-strand-specific uracil glycosylase, dsDNA specific UDG, DsUDG, EC 3.2.2.28, G/U mismatch-specific DNA glycosylase, G:T/U mismatch-specific DNA glycosylase, Mismatch-specific uracil DNA glycosylase, Mug, Xanthine DNA glycosylase |
| Buffer | 20 mM Tris-HCl buffer (pH 8.0), 0.1 M NaCl, 20% glycerol |
| Purity or Quality Grade | >90%, by SDS-PAGE |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction