missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli Biosynthetic arginine decarboxylase Protein

Product Code. p-7155431
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16056561

Brand: enQuireBio™ QP8804ec10ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) Biosynthetic arginine decarboxylase, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_417413.1
Gene ID (Entrez) 947432
Name Biosynthetic arginine decarboxylase Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol speA
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His
Sequence DDMSMGLPSS AGEHGVLRSM QEVAMSSQEA SKMLRTYNIA WWGNNYYDVN ELGHISVCPD PDVPEARVDL AQLVKTREAQ GQRLPALFCF PQILQHRLRS INAAFKRARE SYGYNGDYFL VYPIKVNQHR RVIESLIHSG EPLGLEAGSK AELMAVLAHA GMTRSVIVCN GYKDREYIRL ALIGEKMGHK VYLVIEKMSE IAIVLDEAER LNVVPRLGVR ARLASQGSGK WQSSGGEKSK FGLAATQVLQ LVETLREAGR LDSLQLLHFH LGSQMANIRD IATGVRESAR FYVELHKLGV NIQCFDVGGG LGVDYEGTRS QSDCSVNYGL NEYANNIIWA IGDACEENGL PHPTVITESG RAVTAHHTVL VSNIIGVERN EYTVPTAPAE DAPRALQSMW ETWQEMHEPG TRRSLREWLH DSQMDLHDIH IGYSSGIFSL QERAWAEQLY LSMCHEVQKQ LDPQNRAHRP IIDELQERMA DKMYVNFSLF QSMPDAWGID QLFPVLPLEG LDQVPERRAV LLDITCDSDG AIDHYIDGDG IATTMPMPEY DPENPPMLGF FMVGAYQEIL GNMHNLFGDT EAVDVFVFPD GSVEVELSDE GDTVADMLQY VQLDPKTLLT QFRDQVKKTD LDAELQQQFL EEFEAGLYGY TYLE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.