missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant E. coli beta-lactamase Protein

Product Code. 18611397 Shop All Bio Techne Products
Change view
Click to view available options
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18611397 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18611397 Supplier Novus Biologicals™ Supplier No. NBP2614441mg

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. One unit of enzyme activity is defined as the amount of enzyme which will hydrolyze 1.0 umol of benzyl penicillin in presence of EDTA at pH 7.0 and at 25 C.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number A2T978
For Use With (Application) Enzyme Activity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 100 mM Tris, pH 7.0.
Gene ID (Entrez) 948669
Molecular Weight (g/mol) MolecularWeight-theroretical: 28.9 kDa
Quantity 1 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Gene Alias ampA, AmpC, cephalosporinase, EC 3.5.2.6, ECK4146
Gene Symbol ampC
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Virology Bacteria and Parasites
Purity or Quality Grade >95%, by SDS-PAGE
Protein beta-lactamase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.