missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase Rep Protein

Product Code. 16065851
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16065851

Brand: enQuireBio™ QP8666ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase Rep, was expressed in the hosts and tags indicated.

Specifications

Accession Number WP_001238899.1
Gene ID (Entrez) 948292
Name ATP-dependent DNA helicase Rep Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol rep
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag GST
Sequence MRLNPGQQQA VEFVTGPCLV LAGAGSGKTR VITNKIAHLI RGCGYQARHI AAVTFTNKAA REMKERVGQT LGRKEARGLM ISTFHTLGLD IIKREYAALG MKANFSLFDD TDQLALLKEL TEGLIEDDKV LLQQLISTIS NWKNDLKTPS QAAASAIGER DRIFAHCYGL YDAHLKACNV LDFDDLILLP TLLLQRNEEV RKRWQNKIRY LLVDEYQDTN TSQYELVKLL VGSRARFTVV GDDDQSIYSW RGARPQNLVL LSQDFPALKV IKLEQNYRSS GRILKAANIL IANNPHVFEK RLFSELGYGA ELKVLSANNE EHEAERVTGE LIAHHFVNKT QYKDYAILYR GNHQSRVFEK FLMQNRIPYK ISGGTSFFSR PEIKDLLAYL RVLTNPDDDS AFLRIVNTPK REIGPATLKK LGEWAMTRNK SMFTASFDMG LSQTLSGRGY EALTRFTHWL AEIQRLAERE PIAAVRDLIH GMDYESWLYE TSPSPKAAEM RMKNVNQLFS WMTEMLEGSE LDEPMTLTQV VTRFTLRDMM ERGESEEELD QVQLMTLHAS KGLEFPYVYM VGMEEGFLPH QSSIDEDNID EERRLAYVGI TRAQKELTFT LCKERRQYGE LVRPEPSRFL LELPQDDLIW EQERKVVSAE ERMQKGQSHL ANLKAMMAAK RGK
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.