missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli ATP-dependent DNA helicase RecG Protein

Product Code. 16067410
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16067410

Brand: enQuireBio™ QP711210ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Escherichia coli (strain K12) ATP-dependent DNA helicase RecG, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_418109.1
Gene ID (Entrez) 948162
Name ATP-dependent DNA helicase RecG Protein
Quantity 10 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol recG
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity E. coli
Species E. coli
Protein Tag His-SUMO
Sequence MKGRLLDAVP LSSLTGVGAA LSNKLAKINL HTVQDLLLHL PLRYEDRTHL YPIGELLPGV YATVEGEVLN CNISFGGRRM MTCQISDGSG ILTMRFFNFS AAMKNSLAAG RRVLAYGEAK RGKYGAEMIH PEYRVQGDLS TPELQETLTP VYPTTEGVKQ ATLRKLTDQA LDLLDTCAIE ELLPPELSQG MMTLPEALRT LHRPPPTLQL SDLETGQHPA QRRLILEELL AHNLSMLALR AGAQRFHAQP LSANDTLKNK LLAALPFKPT GAQARVVAEI ERDMALDVPM MRLVQGDVGS GKTLVAALAA LRAIAHGKQV ALMAPTELLA EQHANNFRNW FAPLGIEVGW LAGKQKGKAR LAQQEAIASG QVQMIVGTHA IFQEQVQFNG LALVIIDEQH RFGVHQRLAL WEKGQQQGFH PHQLIMTATP IPRTLAMTAY ADLDTSVIDE LPPGRTPVTT VAIPDTRRTD IIDRVHHACI TEGRQAYWVC TLIEESELLE AQAAEATWEE LKLALPELNV GLVHGRMKPA EKQAVMASFK QGELHLLVAT TVIEVGVDVP NASLMIIENP ERLGLAQLHQ LRGRVGRGAV ASHCVLLYKT PLSKTAQIRL QVLRDSNDGF VIAQKDLEIR GPGELLGTRQ TGNAEFKVAD LLRDQAMIPE VQRLARHIHE RYPQQAKALI ERWMPETERY SNA
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.