missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli ACKA Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | ACKA Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | E. coli |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMSSKLVL VLNCGSSSLK FAIIDAVNGE EYLSGLAECF HLPEARIKWK MDGNKQEAAL GAGAAHSEAL NFIVNTILAQ KPELSAQLTA IGHRIVHGGE KYTSSVVIDE SVIQGIKDAA SFAPLHNPAH LIGIEEALKS FPQLKDKNVA VFDTAFHQTM PEESYLYALP YNLYKEHGIR RYGAHGTSHF YVTQEAAKML NKPVEELNII TCHLGNGGSV SAIRNGKCVD TSMGLTPLEG LVMGTRSGDI DPAIIFHLHD TLGMSVDAIN KLLTKESGLL GLTEVTSDCR YVEDNYATKE DAKRAMDVYC HRLAKYIGAY TALMDGRLDA VVFTGGIGEN AAMVRELSLG KLGVLGFEVD HERNLAARFG KSGFINKEGT RPAVVIPTNE ELVIAQDASR LTA |
| Buffer | ACKA protein (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction