missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant E. coli 50S ribosomal protein L36 Protein

A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L36, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8625-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16075641

  • £2480.00 / 1mg
Estimated Shipment: 15-01-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_417758.1
50S ribosomal protein L36 Protein
Research Use Only
rpmJ
Recombinant Protein
E. coli
MKVRASVKKL CRNCKIVKRD GVIRVICSAE PKHKQRQG
Greater than 90% as determined by SDS-PAGE.
32366016
1 mg
Not determined.
Full Length
E. coli
GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers