missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 50S ribosomal protein L29 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 50S ribosomal protein L29, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8634-ec-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417771.1 | |
50S ribosomal protein L29 Protein | |
Research Use Only | |
rpmC | |
Recombinant Protein | |
E. coli | |
MKAKELREKS VEELNTELLN LLREQFNLRM QAASGQLQQS HLLKQVRRDV ARVKTLLNEK AGA | |
Greater than 90% as determined by SDS-PAGE. |
947807 | |
200 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |