missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli 30S ribosomal protein S19 Protein
A DNA sequence encoding the Escherichia coli (strain K12) 30S ribosomal protein S19, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8617-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_417775.1 | |
30S ribosomal protein S19 Protein | |
Research Use Only | |
rpsS | |
Recombinant Protein | |
E. coli | |
PRSLKKGPFI DLHLLKKVEK AVESGDKKPL RTWSRRSTIF PNMIGLTIAV HNGRQHVPVF VTDEMVGHKL GEFAPTRTYR GHAADKKAKK K | |
Greater than 90% as determined by SDS-PAGE. |
31655999 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |