missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dog Serum Albumin / HSA / ALB Protein

Product Code. 15906649
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 15906649

Brand: enQuireBio™ QP5642ec200ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

A DNA sequence encoding the Canis lupus familiaris (Dog) (Canis familiaris) Serum Albumin / HSA / ALB, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_001003026.1
Gene ID (Entrez) 403550
Name Serum Albumin / HSA / ALB Protein
Quantity 200 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol ALB
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Dog
Species E. coli
Protein Tag His
Sequence EAYKSEIAHR YNDLGEEHFR GLVLVAFSQY LQQCPFEDHV KLAKEVTEFA KACAAEESGA NCDKSLHTLF GDKLCTVASL RDKYGDMADC CEKQEPDRNE CFLAHKDDNP GFPPLVAPEP DALCAAFQDN EQLFLGKYLY EIARRHPYFY APELLYYAQQ YKGVFAECCQ AADKAACLGP KIEALREKVL LSSAKERFKC ASLQKFGDRA FKAWSVARLS QRFPKADFAE ISKVVTDLTK VHKECCHGDL LECADDRADL AKYMCENQDS ISTKLKECCD KPVLEKSQCL AEVERDELPG DLPSLAADFV EDKEVCKNYQ EAKDVFLGTF LYEYARRHPE YSVSLLLRLA KEYEATLEKC CATDDPPTCY AKVLDEFKPL VDEPQNLVKT NCELFEKLGE YGFQNALLVR YTKKAPQVST PTLVEVSRKL GKVGTKCCKK PESERMSCAE DFLSVVLNRL CVLHEKTPVS ERVTKCCSES LVNRRPCFSG LEVDETYVPK EFNAETFTFH ADLCTLPEAE KQVKKQTALV ELLKHKPKAT DEQLKTVMGD FGAFVEKCCA AENKEGCFSE EGPKLVAAAQ AALV
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.