missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Denis GH Protein

A cDNA sequence encoding the GH was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11961-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15938938

  • £2315.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

GH Protein
Research Use Only
Recombinant Protein
E. coli
MDRVVLMLSV MSLGVSSQPI TDGQRLFSIA VSRVQHLHLL AQRLFSDFES SLQTEEQRQL NKIFLQDFCN SDYIISPIDK HETQRSSVLK LLSISYRLVE SWEFPSRSLS GGSAPRNQIS PKLSELKTGI HLLIRANEDG AEIFPDSSAL QLAPYGNYYQ SLGTDESLRR TYELLACFKK DMHKVETYLT VAKCRLSPEA NCTL
Greater than 97.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Denis
Untagged
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.0045mM NaHCO3.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers