missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus, ST3 Dengue NS1 Protein
A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10496-100 ug
Additional Details : Weight : 0.01000kg
Specifications
Dengue Virus, ST3 Dengue NS1 Protein | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus, ST3 | |
His |
Protein is >90% pure as determined by 12% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphate buffered saline and 50mM arginine. |