missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Polyvalent ELISA Protein
A cDNA sequence encoding the Dengue Polyvalent ELISA was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11651-1mg
This item is not returnable.
View return policy
Specifications
Dengue Polyvalent ELISA Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
50mM sodium chloride-phosphorous buffer, pH 7.4. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
His | |
Protein is >95% pure as determined by 10% PAGE (coomassie staining). |