missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Polyvalent ELISA Protein

A cDNA sequence encoding the Dengue Polyvalent ELISA was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11651-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15967918

  • £170.83 / 100µg
Estimated Shipment: 24-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Dengue Polyvalent ELISA Protein
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 10% PAGE (coomassie staining).
Research Use Only
Recombinant Protein
E. coli
50mM sodium chloride-phosphorous buffer, pH 7.4.
Videos
SDS
Documents
Certificates
Special Offers

Special Offers