missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Envelope-3 45kDa Protein

A cDNA sequence encoding the Dengue Envelope-3 45kDa was constructed and used to recombinantly synthesize the protein.

£170.83 - £1324.47

Specifications

Name Dengue Envelope-3 45kDa Protein
Product Type Recombinant Protein
Protein Tag His
Host Species E. coli
Cross Reactivity Dengue Virus
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15937868
View Documents Promotion Details
enQuireBio™
QP11634-100UG
100 μg
£170.83
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15957868
View Documents Promotion Details
enQuireBio™
QP11634-500UG
500 μg
£662.78
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
15947868
View Documents Promotion Details
enQuireBio™
QP11634-1MG
1 mg
£1324.47
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 24-02-2022
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Dengue Envelope-3 45kDa Protein
His
Dengue Virus
Recombinant Protein
E. coli
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
SDS
Product Certifications