missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-2 22kDa Protein
A cDNA sequence encoding the Dengue Envelope-2 22kDa was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11628-1mg
Additional Details : Weight : 0.01000kg
Specifications
Dengue Envelope-2 22kDa Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
Phosphorous buffered saline, pH 7.4. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |