missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus Dengue Envelope-1 & 4 Protein

A cDNA sequence encoding the Dengue Envelope-1 & 4 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP11621-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15957828

  • £1320.00 / 1mg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Dengue Envelope-1 & 4 Protein
Research Use Only
Recombinant Protein
E. coli
Phosphate buffered saline, pH 7.4.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Dengue Virus
His
Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers