missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-1 & 3 Protein
A cDNA sequence encoding the Dengue Envelope-1 & 3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11620-1mg
Additional Details : Weight : 0.01000kg
Specifications
Dengue Envelope-1 & 3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
The protein solution contains Phosphate buffered saline, pH 7.4. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
His | |
Protein is >95% pure as determined by 12% PAGE (coomassie staining). |