missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Cytomegalovirus Cytomegalovirus gH Pentamer Complex VR1814 His (C-Term) Protein
Shop All Bio Techne ProductsSpecifications
Specifications
| For Use With (Application) | SDS-PAGE |
| Formulation | Dulbecco's PBS, 0.2 um filter sterilised. |
| Quantity | 0.5 mg |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | envelope glycoprotein H |
| Purity or Quality Grade | >95% pure by SDS-PAGE |
| Protein | Cytomegalovirus gH Pentamer Complex |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?