missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Cynomolgus/Rhesus Macaque IL-17F His Protein

Product Code. 18795936
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18795936 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18795936 Supplier R&D Systems™ Supplier No. 10225IL050
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Measured by its ability to induce IL-6 secretion by NIH-3T3 mouse embryonic fibroblast cells.

The ED50 for this effect is 5 to 40 ng/mL
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_001248216.1
For Use With (Application) Bioactivity
Gene ID (Entrez) 112744
Molecular Weight (g/mol) 17 to 25 kDa, under reducing conditions
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived IL-17F protein Arg31-Gln163, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration < 0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Species Cynomolgus Monkey, Rhesus macaque
Reconstitution Reconstitute at 200 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Product Line R&D Systems
Purity or Quality Grade > 95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.