missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus/Rhesus Macaque CD27 Fc Protein
Description
Measured by its ability to inhibit CD70-induced IL-8 secretion in HT1080 human fibrosarcoma cells transfected with human CD27. The ED50 for this effect is 0.25-1.5 μg/mL.
Specifications
Specifications
| Accession Number | XP_005569963.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS. |
| Gene ID (Entrez) | 939 |
| Molecular Weight (g/mol) | M.W. (Observed): 61-68 kDa, reducing conditions; M.W. (theoretical): 46 kDa |
| Quantity | 100 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived CD27/TNFRSF7 protein Cynomolgus Monkey CD27 (Ala20-Arg191) (N-terminus) Accession # XP_005569963.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, <= -20 °C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?